Assay Details
| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037331.3 | NP_001032408.1 | 2 - 3 | 406 | 120 | |
| GenBank mRNA | AK093452.1 | - | 2 - 3 | 406 | 120 | |
| BC107486.1 | - | 2 - 3 | 337 | 40032395 | 120 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000382638.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000579022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | ENST00000584508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001037331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009058.hg.1 | NM_001303482.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000578340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007702.hg.1 | ENST00000579386.2 | Ensembl |

SMIM21
Genomic Map