Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011515256.2 | XP_011513558.1 | 1 - 2 | 62 | 87 | |
| XM_011515257.2 | XP_011513559.1 | 1 - 2 | 62 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011515256.2, XM_011515257.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |

KBTBD2
Genomic Map