Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015483.2 | NP_056298.2 | 3 - 4 | 992 | 127 | |
| XM_005249696.2 | XP_005249753.1 | 3 - 4 | 848 | 127 | ||
| XM_005249697.2 | XP_005249754.1 | 3 - 4 | 581 | 127 | ||
| XM_011515256.2 | XP_011513558.1 | 3 - 4 | 727 | 127 | ||
| XM_011515257.2 | XP_011513559.1 | 3 - 4 | 461 | 127 | ||
| GenBank mRNA | AF151831.1 | - | 2 - 3 | 401 | 127 | |
| AK292802.1 | - | 3 - 4 | 714 | 127 | ||
| BC037887.2 | - | 3 - 4 | 896 | 5274706 | 127 | |
| BC047107.1 | - | 3 - 4 | 729 | 5275217 | 127 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000304056.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000423022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000424468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000452926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000453627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000477129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000485611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | ENST00000621876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010687.hg.1 | NM_015483.2 | Refseq |

KBTBD2
Genomic Map