Assay Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005249789.1 | XP_005249846.1 | 1 - 2 | 256 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005249789.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396667.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000396668.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000420833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000442107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000444443.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000453686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000462754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | ENST00000492762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_001134232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006727.hg.1 | NM_018374.3 | Refseq |

TMEM106B
Genomic Map