| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000407277.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000407277.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000407277.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000498264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
ENST00000498264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000342526.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000407277.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000407277.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000407277.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000418075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000498264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
ENST00000498264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010334.hg.1 |
NM_001004320.1 |
Refseq |