Assay Details
| Gene Symbol: AGMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006715730.1 | XP_006715793.1 | 12 - 13 | 1430 | 139 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000407277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000407277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000407277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000498264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | ENST00000498264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000342526.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000407277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000407277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000407277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000418075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000498264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | ENST00000498264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010334.hg.1 | NM_001004320.1 | Refseq |

AGMO
Genomic Map