Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250482.3 | XP_005250539.1 | 1 - 2 | 97 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250482.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000341567.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000413593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000418375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000424964.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000433271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000484751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | ENST00000607045.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007857.hg.1 | NM_017994.4 | Refseq |

TMEM248
Genomic Map