Assay Details
| Gene Symbol: ZNF736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170905.2 | NP_001164376.1 | 4 | 8124 | 92 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006969.hg.1 | ENST00000621243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006969.hg.1 | ENST00000621243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006971.hg.1 | ENST00000614118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006971.hg.1 | ENST00000614118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007756.hg.1 | ENST00000415181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007756.hg.1 | ENST00000415181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007756.hg.1 | ENST00000415181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007251.hg.1 | ENST00000616864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007251.hg.1 | ENST00000616864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000441518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000441518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000492322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000492322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000492322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000606716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000606716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | ENST00000606716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006779.hg.1 | ENST00000438459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006779.hg.1 | ENST00000438459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007510.hg.1 | ENST00000509929.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007510.hg.1 | ENST00000509929.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011189.hg.1 | ENST00000444048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011189.hg.1 | ENST00000444048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006811.hg.1 | ENST00000445313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006811.hg.1 | ENST00000445313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007756.hg.1 | ENST00000415181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007756.hg.1 | ENST00000415181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007756.hg.1 | ENST00000415181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007251.hg.1 | ENST00000616864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007251.hg.1 | ENST00000616864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007510.hg.1 | ENST00000509929.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007510.hg.1 | ENST00000509929.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006811.hg.1 | ENST00000445313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006811.hg.1 | ENST00000445313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011189.hg.1 | ENST00000444048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011189.hg.1 | ENST00000444048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006779.hg.1 | ENST00000438459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006779.hg.1 | ENST00000438459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006969.hg.1 | ENST00000621243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006969.hg.1 | ENST00000621243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000355095.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000423484.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000438373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000441518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000441518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000465343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000488621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000492322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000492322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000492322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000493036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000606716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000606716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | ENST00000606716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001170905.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013368.hg.1 | NM_001294255.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006971.hg.1 | ENST00000614118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006971.hg.1 | ENST00000614118.1 | Ensembl |

ZNF736
Genomic Map