Assay Details
| Gene Symbol: LOC283299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036678.1 | 5 - 6 | 769 | 70 | ||
| GenBank mRNA | AK091855.1 | - | 5 - 6 | 769 | 70 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000502624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000502624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000502624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000502624.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000527847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000529488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | ENST00000533634.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013142.hg.1 | NR_036678.1 | Refseq |

LOC283299
Genomic Map