Assay Details
| Gene Symbol: BCL9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271511.2 | XP_005271568.1 | 1 - 2 | 835 | 105 | |
| XM_005271512.2 | XP_005271569.1 | 1 - 2 | 387 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005271511.2, XM_005271512.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of epithelial to mesenchymal transition
regulation of cell morphogenesis
negative regulation of transforming growth factor beta receptor signaling pathway
somatic stem cell population maintenance
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
transcription coactivator activity
beta-catenin binding
transcription, DNA-templated
positive regulation of epithelial to mesenchymal transition
regulation of cell morphogenesis
negative regulation of transforming growth factor beta receptor signaling pathway
somatic stem cell population maintenance
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
transcription coactivator activity
beta-catenin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000530293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000530293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | ENST00000530293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000334801.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000527266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000530293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000530293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | ENST00000530293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013230.hg.1 | NM_182557.2 | Refseq |

BCL9L
Genomic Map