Assay Details
| Gene Symbol: TARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S60779.1 | - | 1 | 124 | 99 | |
| X74774.1 | - | 1 | 124 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S60779.1 | - | 1 | 124 | 99 | |
| X74774.1 | - | 1 | 124 | 100 | ||
Set Membership: |
|