Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136044.1 | NP_001129516.1 | 3 | 1288 | 116 | |
| NM_031434.3 | NP_113622.1 | 3 | 1751 | 116 | ||
| GenBank mRNA | AY037155.1 | - | 3 | 1269 | 116 | |
| AY358481.1 | - | 3 | 1247 | 116 | ||
| AY603380.1 | - | 2 | 1273 | 116 | ||
| BC000936.1 | - | 3 | 1288 | 3449979 | 116 | |
| BC033182.1 | - | 3 | 1426 | 4586932 | 116 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |

TMUB1
Genomic Map