Assay Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC034542.1 | - | 1 | 721 | 81 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009164.hg.1 | ENST00000510610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009164.hg.1 | ENST00000510610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009164.hg.1 | ENST00000510610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009164.hg.1 | ENST00000510610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009164.hg.1 | ENST00000510610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009164.hg.1 | ENST00000510610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000301263.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | ENST00000518884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | NM_003695.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012088.hg.1 | NM_003695.2 | Refseq |

LY6D
Genomic Map