Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003012.4 | NP_003003.3 | 3 | 4057 | 112 | |
| GenBank mRNA | AF056087.1 | - | 3 | 4051 | 112 | |
| AK127331.1 | - | 3 | 3469 | 112 | ||
| AL049995.1 | - | 1 | 119 | 112 | ||
| BC036503.1 | - | 3 | 4063 | 5265277 | 112 | |
| BQ002015.1 | - | 1 | 112 | |||
More Information
Set Membership: |
|

SFRP1
Genomic Map