Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306188.1 | NP_001293117.1 | 4 - 5 | 711 | 197 | |
| XM_017029654.1 | XP_016885143.1 | 4 - 5 | 665 | 197 | ||
| GenBank mRNA | AK299172.1 | - | 3 - 4 | 501 | 197 | |
| CB997365.1 | - | 4 - 5 | 619 | 197 | ||
More Information
Set Membership: |
|

MOSPD1
Genomic Map