| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000548348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000548348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000548348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000432328.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000546455.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000548348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000548348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000548348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000549630.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
ENST00000551777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_001281429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007461.hg.1 |
NM_138371.2 |
Refseq |