Assay Details
| Gene Symbol: C12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537896.2 | XP_011536198.1 | 3 - 4 | 754 | 99 | |
| XM_017018796.1 | XP_016874285.1 | 3 - 4 | 768 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011537896.2, XM_017018796.1 transcripts.
Set Membership: |
|

C12orf54
Genomic Map