Assay Details
| Gene Symbol: LINC00459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX113955.1 | - | 1 - 2 | 91 | 87 | |
| R46019.1 | - | 1 - 2 | 92 | 87 | ||
| T24087.1 | - | 1 - 2 | 80 | 87 | ||
| Z39313.1 | - | 1 - 2 | 87 | |||
More Information
Set Membership: |
|

LINC00459
Genomic Map