Assay Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020645.1 | XP_016876134.1 | 1 - 2 | 87 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017020645.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000365477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000491647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000491647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000491647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000332592.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000365477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000375403.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000438545.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000439909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000460318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000465938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000478244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000482038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000491647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000491647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000491647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | ENST00000496873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009907.hg.1 | NM_001014283.1 | Refseq |

DCUN1D2
Genomic Map