Assay Details
| Gene Symbol: SACS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278055.1 | NP_001264984.1 | 4 - 5 | 540 | 81 | |
| NM_014363.5 | NP_055178.3 | 6 - 7 | 1071 | 81 | ||
| XM_005266338.2 | XP_005266395.1 | 6 - 7 | 1071 | 81 | ||
| XM_011535039.2 | XP_011533341.1 | 5 - 6 | 679 | 81 | ||
| XM_017020539.1 | XP_016876028.1 | 5 - 6 | 679 | 81 | ||
| GenBank mRNA | AK125458.1 | - | 4 - 5 | 540 | 81 | |
| AW295072.1 | - | 3 - 4 | 295 | 81 | ||
| BX640926.1 | - | 2 - 3 | 154 | 81 | ||
| CR987846.1 | - | 3 - 4 | 288 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278055.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000476776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000476776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | ENST00000476776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000382298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000402364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000423156.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000455470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000476776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000476776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | ENST00000476776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_001278055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008329.hg.1 | NM_014363.5 | Refseq |

SACS
Genomic Map