Assay Details
| Gene Symbol: CGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015433.1 | - | 6 - 7 | 501 | 96 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |

CGNL1
Genomic Map