| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000457093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000457093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000457093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000457093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
ENST00000621082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008845.hg.1 |
NM_001207020.2 |
Refseq |