Assay Details
| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005577.2 | NP_005568.2 | 34 - 35 | 5518 | 70 | |
| GenBank mRNA | X06290.1 | - | 12967 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005577.2 | NP_005568.2 | 34 - 35 | 5518 | 70 | |
| GenBank mRNA | X06290.1 | - | 12967 | 70 | ||
Set Membership: |
|