Assay Details
| Gene Symbol: OR1J4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004452.1 | NP_001004452.1 | 1 | 589 | 71 | |
| GenBank mRNA | BC131606.1 | - | 1 | 588 | 71 | |
| X64979.1 | - | 1 | 215 | 71 | ||
More Information
Set Membership: |
|

OR1J4
Genomic Map