Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195214.1 | NP_001182143.1 | 6 | 4364 | 78 | |
| NM_173834.3 | NP_776195.2 | 7 | 4493 | 78 | ||
| XM_011530924.1 | XP_011529226.1 | 7 | 4275 | 78 | ||
| GenBank mRNA | AL117461.1 | - | 1 | 2187 | 78 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |

YIPF6
Genomic Map