| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000601887.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
ENST00000601887.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000358067.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000500161.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000597186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000598312.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000601887.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
ENST00000601887.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011606.hg.1 |
NM_024620.3 |
Refseq |