Assay Details
| Gene Symbol: ZNF329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095349.1 | - | 4 - 5 | 670 | 78 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010361.hg.1 | ENST00000585314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010361.hg.1 | ENST00000585314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |

ZNF329
Genomic Map