| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000590655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000590655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000590655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000269973.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000588432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000589703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000590655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000590655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000590655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
ENST00000615985.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010873.hg.1 |
NM_003425.3 |
Refseq |