Assay Details
| Gene Symbol: ANKRD26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017015928.1 | XP_016871417.1 | 38 - 39 | 6878 | 114 | |
| XM_017015929.1 | XP_016871418.1 | 39 - 40 | 6866 | 114 | ||
More Information
Set Membership: |
|

ANKRD26
Genomic Map