Assay Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 2353 | 113 | |
| XM_005245848.3 | XP_005245905.1 | 7 | 2588 | 113 | ||
| XM_005245849.3 | XP_005245906.1 | 7 | 2693 | 113 | ||
| XM_005245850.3 | XP_005245907.1 | 8 | 2713 | 113 | ||
| XM_005245851.3 | XP_005245908.1 | 7 | 2585 | 113 | ||
| XM_005245852.3 | XP_005245909.1 | 6 | 2362 | 113 | ||
| XM_011541255.1 | XP_011539557.1 | 7 | 2267 | 113 | ||
| XM_011541256.2 | XP_011539558.1 | 7 | 2707 | 113 | ||
| XM_011541257.2 | XP_011539559.1 | 6 | 3462 | 113 | ||
More Information
Set Membership: |
|

AHDC1
Genomic Map