| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006612.hg.1 |
ENST00000587737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006612.hg.1 |
ENST00000587737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006612.hg.1 |
ENST00000587737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000589201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000589201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
ENST00000589201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006612.hg.1 |
ENST00000587737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006612.hg.1 |
ENST00000587737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006612.hg.1 |
ENST00000587737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000379053.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000589201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000589201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
ENST00000589201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009980.hg.1 |
NR_015406.2 |
Refseq |