| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
ENST00000309880.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008013.hg.1 |
NM_145719.2 |
Refseq |