Assay Details
| Gene Symbol: CLIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317009.1 | NP_001303938.1 | 7 | 110 | ||
| NM_053277.2 | NP_444507.1 | 6 | 110 | |||
| GenBank mRNA | AF308293.1 | - | 1 | 415 | 110 | |
| AF448438.1 | - | 6 | 110 | |||
| AF448439.1 | - | 7 | 110 | |||
| AK092733.1 | - | 5 | 110 | |||
| BC040196.2 | - | 6 | 110 | |||
| CA442589.1 | - | 1 | 454 | 110 | ||
More Information
Set Membership: |
|

CLIC6
Genomic Map