Assay Details
| Gene Symbol: C22orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093880.1 | - | 3 | 754 | 126 | |
| BC113450.1 | - | 1 | 116 | 8322502 | 126 | |
| BC113452.1 | - | 1 | 116 | 8322504 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC113450.1, BC113452.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |

C22orf15
Genomic Map