Assay Details
| Gene Symbol: USP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK303463.1 | - | 7 - 8 | 828 | 113 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006546.hg.1 | ENST00000413504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006546.hg.1 | ENST00000413504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006546.hg.1 | ENST00000622906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006546.hg.1 | ENST00000622906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008076.hg.1 | ENST00000417463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008076.hg.1 | ENST00000417463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000429995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000429995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000429995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000451267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000451267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000614584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000614584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000620909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000620909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007929.hg.1 | ENST00000620909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006697.hg.1 | ENST00000434730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006697.hg.1 | ENST00000434730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000292729.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000454608.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000470202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008036.hg.1 | ENST00000486536.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000429995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000429995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000429995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000451267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000451267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000614584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000614584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000620909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000620909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007929.hg.1 | ENST00000620909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008076.hg.1 | ENST00000417463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008076.hg.1 | ENST00000417463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006546.hg.1 | ENST00000413504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006546.hg.1 | ENST00000413504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006546.hg.1 | ENST00000622906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006546.hg.1 | ENST00000622906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006697.hg.1 | ENST00000434730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006697.hg.1 | ENST00000434730.1 | Ensembl |

USP41
Genomic Map