Assay Details
Gene Symbol: LRPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002337.3 | NP_002328.1 | 8 | 1459 | 107 | |
NR_110005.1 | 8 | 1337 | 107 | |||
GenBank mRNA | AK222504.1 | - | 8 | 1384 | 107 | |
AK222527.1 | - | 8 | 1383 | 107 | ||
BX647984.1 | - | 8 | 1384 | 107 | ||
M63959.1 | - | 8 | 1387 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
receptor-mediated endocytosis
negative regulation of very-low-density lipoprotein particle clearance
vesicle-mediated transport
negative regulation of protein binding
extracellular negative regulation of signal transduction
negative regulation of beta-amyloid clearance
asialoglycoprotein receptor activity
protein binding
heparin binding
lipase binding
receptor antagonist activity
low-density lipoprotein particle receptor binding
unfolded protein binding
very-low-density lipoprotein particle receptor binding
protein folding
receptor-mediated endocytosis
negative regulation of very-low-density lipoprotein particle clearance
vesicle-mediated transport
negative regulation of protein binding
extracellular negative regulation of signal transduction
negative regulation of beta-amyloid clearance
asialoglycoprotein receptor activity
protein binding
heparin binding
lipase binding
receptor antagonist activity
low-density lipoprotein particle receptor binding
unfolded protein binding
very-low-density lipoprotein particle receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | ENST00000564650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000296325.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000500728.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000509198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000515119.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | ENST00000564650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NM_002337.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009818.hg.1 | NR_110005.1 | Refseq |
