| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
ENST00000371268.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008414.hg.1 |
NM_182532.2 |
Refseq |