Assay Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022347.3 | NP_071742.1 | 3 | 673 | 90 | |
| XM_017000477.1 | XP_016855966.1 | 2 | 1346 | 90 | ||
| XM_017000478.1 | XP_016855967.1 | 2 | 679 | 90 | ||
| GenBank mRNA | AK289793.1 | - | 3 | 648 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK289793.1, NM_022347.3, XM_017000477.1, XM_017000478.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007857.hg.1 | ENST00000631318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007857.hg.1 | ENST00000631318.1 | Ensembl |

TOR1AIP2
Genomic Map