| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000256151.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000547758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000548126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000550589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
ENST00000552606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NM_014167.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011400.hg.1 |
NR_033192.1 |
Refseq |