Assay Details
| Gene Symbol: PLAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182832.2 | NP_878252.2 | 1 | 4771 | 96 | |
| GenBank mRNA | AF269287.1 | - | 1 | 1020 | 96 | |
| AK027868.1 | - | 1 | 2433 | 96 | ||
| L13197.1 | - | 1 | 447 | 96 | ||
| Gene Symbol: BACE2-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI189527.1 | - | 2 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF269287.1, NM_182832.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007200.hg.1 | ENST00000433378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007200.hg.1 | ENST00000433378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007200.hg.1 | ENST00000433378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007200.hg.1 | ENST00000433378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000414699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000430327.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000440221.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000536486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000623119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | ENST00000624999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008200.hg.1 | NM_182832.2 | Refseq |

PLAC4
Genomic Map