Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019058.2 | NP_061931.1 | 3 | 1569 | 105 | |
| GenBank mRNA | AF335324.1 | - | 3 | 1585 | 105 | |
| AK000507.1 | - | 3 | 1564 | 105 | ||
| AL136668.1 | - | 3 | 1568 | 105 | ||
| BC000708.1 | - | 3 | 1475 | 3507244 | 105 | |
| BC007714.1 | - | 3 | 1557 | 4302878 | 105 | |
| BC015236.1 | - | 3 | 1543 | 3906374 | 105 | |
| CN428398.1 | - | 1 | 185 | 105 | ||
More Information
Set Membership: |
|

DDIT4
Genomic Map