| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
ENST00000258400.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016006.hg.1 |
NM_000867.4 |
Refseq |