Assay Details
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293289.1 | NP_001280218.1 | 1 | 215 | 65 | |
| NM_001293290.1 | NP_001280219.1 | 1 | 215 | 65 | ||
| XM_011537685.2 | XP_011535987.1 | 1 | 626 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293289.1, NM_001293290.1, XM_011537685.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000344120.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000434127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000503582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000509364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | ENST00000511815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001127496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293289.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_001293290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012312.hg.1 | NM_030964.3 | Refseq |

SPRY4
Genomic Map