Assay Details
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017024243.1 | XP_016879732.1 | 12 | 1700 | 60 | |
| GenBank mRNA | AK125672.1 | - | 2 | 1524 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017024243.1 | XP_016879732.1 | 12 | 1700 | 60 | |
| GenBank mRNA | AK125672.1 | - | 2 | 1524 | 60 | |
Set Membership: |
|