| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010784.hg.1 |
ENST00000390345.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010784.hg.1 |
ENST00000390345.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010788.hg.1 |
ENST00000426402.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010788.hg.1 |
ENST00000426402.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010788.hg.1 |
ENST00000426402.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010788.hg.1 |
ENST00000426402.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010784.hg.1 |
ENST00000390345.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010784.hg.1 |
ENST00000390345.2 |
Ensembl |