Assay Details
| Gene Symbol: HIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ550616.1 | - | 1 | 146 | 74 | |
| AJ583693.1 | - | 1 | 74 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ550616.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000322941.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000399849.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000571990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000574370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000576444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | ENST00000619757.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_001098202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006535.hg.1 | NM_006497.3 | Refseq |

HIC1
Genomic Map