Assay Details
| Gene Symbol: MAFIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046439.2 | 5 | 1682 | 84 | ||
| GenBank mRNA | AK074146.1 | - | 2 | 2135 | 84 | |
| AK096952.1 | - | 5 | 1668 | 84 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007534.hg.1 | NR_038329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000400754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | ENST00000613230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046440.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC14_GL000194v1_random00006432.hg.1 | NR_046442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000416067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | ENST00000559466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038328.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007534.hg.1 | NR_038329.2 | Refseq |

MAFIP
Genomic Map