Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181789.2 | NP_861454.2 | 1 | 120 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_181789.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000335449.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000396399.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000464150.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000558426.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000559317.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000560690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000561194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | ENST00000612989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007231.hg.1 | NM_181789.3 | Refseq |

GLDN
Genomic Map