Assay Details
| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297436.1 | NP_001284365.1 | 5 | 1614 | 96 | |
| NM_001523.3 | NP_001514.2 | 5 | 1617 | 96 | ||
| XM_011526884.2 | XP_011525186.1 | 4 | 1468 | 96 | ||
| GenBank mRNA | BC035837.1 | - | 5 | 1596 | 5589083 | 96 |
| D84424.1 | - | 4 | 1601 | 96 | ||
| U59269.1 | - | 5 | 1593 | 96 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycosaminoglycan biosynthetic process
cell adhesion
negative regulation of fibroblast migration
hyaluronan biosynthetic process
cellular response to platelet-derived growth factor stimulus
estrous cycle
extracellular polysaccharide biosynthetic process
extracellular matrix assembly
hyaluronan synthase activity
glycosaminoglycan biosynthetic process
cell adhesion
negative regulation of fibroblast migration
hyaluronan biosynthetic process
cellular response to platelet-derived growth factor stimulus
estrous cycle
extracellular polysaccharide biosynthetic process
extracellular matrix assembly
hyaluronan synthase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000222115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000540069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000594621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | ENST00000601714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001297436.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011270.hg.1 | NM_001523.3 | Refseq |

HAS1
Genomic Map