| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009587.hg.1 |
ENST00000508231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009587.hg.1 |
ENST00000508231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008506.hg.1 |
ENST00000553003.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000340385.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000460950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
ENST00000470017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010648.hg.1 |
NM_004905.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009587.hg.1 |
ENST00000508231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009587.hg.1 |
ENST00000508231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008506.hg.1 |
ENST00000553003.1 |
Ensembl |